Learn More
Uline Amber Glass Jar 32oz
Catalog No. NC1488071
Encompass
Store tablets, powders and light-sensitive materials.Wide mouth for easy access to contents.Phenolic black caps included.FDA compliant.
Catalog No. NC1488071 Supplier Uline Supplier No. S20593CS
Store tablets, powders and light-sensitive materials.Wide mouth for easy access to contents.Phenolic black caps included.FDA compliant.