Learn More
Tewes Corporation AMBER ZIPPER BAGS 4X6 3 MIL
SDP
Catalog No. NC1841708
Encompass_Preferred
Reclosable Amber Bags offer translucent UV protection for light sensitive medications or electronic parts during transport and storage. Bags meet USP XXII and USP XXIII procedure 66.1 for light transmission not exceeding 10% at any wavelength from 290nm to 450nm. 1000/CASE.
Catalog No. NC1841708 Supplier Tewes Corporation Supplier No. ZAB46
Reclosable Amber Bags offer translucent UV protection for light sensitive medications or electronic parts during transport and storage. Bags meet USP XXII and USP XXIII procedure 66.1 for light transmission not exceeding 10% at any wavelength from 290nm to 450nm. 1000/CASE.