Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AmberChrom™, 1x8, 100-200 mesh, Cl-form, Thermo Scientific™

Catalog No. AC472961000
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC472961000 100 g
AC472965000 500 g
AC472960025 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. AC472961000 Supplier Thermo Scientific Chemicals Supplier No. 472961000
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 69011-19-4 , 7732-18-5
Molecular Formula C31H39N
Molecular Weight (g/mol) 0.00
MDL Number MFCD00146763
InChI Key HADXLRSCRPYPJJ-UHFFFAOYSA-N
IUPAC Name 1,2-diethenylbenzene; 1-ethenyl-2-ethylbenzene; ethenylbenzene; trimethylamine
SMILES *

Specifications

Physical Form Beads
CAS Min % 35%, 55%
CAS Max % 45%, 65%
Ion Exchange Capacity ≥1.2 eq/L (Cl form)
Particle Size 100 to 200 mesh, ≤15 % (< 88 μm), ≤15 % (> 250 μm)
Packaging Plastic Bottle
Water Retention Capacity 39 to 45 % (Cl form)
Color White to Yellow
Quantity 100 g
Solubility Information Insoluble in water
Chemical Name or Material AmberChrom™
Show More Show Less
Recommended Storage Normal conditions
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.