Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AmberChrom™, 1x8, 50-100 mesh, Cl-form, Thermo Scientific™

Catalog No. AC472955000
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC472955000 500 g
AC472951000 100 g
AC472950025 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. AC472955000 Supplier Thermo Scientific Chemicals Supplier No. 472955000
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 69011-19-4 , 7732-18-5
Molecular Formula C31H39N
Molecular Weight (g/mol) 0.00
MDL Number MFCD00146763
InChI Key HADXLRSCRPYPJJ-UHFFFAOYSA-N
IUPAC Name 1,2-diethenylbenzene; 1-ethenyl-2-ethylbenzene; ethenylbenzene; trimethylamine
SMILES *

Specifications

Physical Form Beads
CAS Min % 35%, 55%
CAS Max % 45%, 65%
Ion Exchange Capacity ≥1.2 eq/L (Cl form)
Particle Size 50 to 100 mesh, ≤15 % (< 180 μm), ≤15 % (> 500 μm)
Packaging Plastic Bottle
Water Retention Capacity 43 to 48 % (Cl form)
Color Tan to Yellow to White
Quantity 500 g
Solubility Information Insoluble in water
Chemical Name or Material AmberChrom™
Show More Show Less
Recommended Storage Normal conditions
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.