Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AmberChrom™, 50WX8, 100-200 mesh, H-form, Thermo Scientific™

Catalog No. AC472920025
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC472920025 2.5 kg
AC472921000 100 g
AC472925000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. AC472920025 Supplier Thermo Scientific Chemicals Supplier No. 472920025
  • $2,210.00
    $1,869.65 / Each of 1

    Save $340.35

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 69011-20-7 , 7732-18-5
Molecular Formula C28H30
Molecular Weight (g/mol) 0.00
MDL Number MFCD00132722
InChI Key NWUYHJFMYQTDRP-UHFFFAOYSA-N
IUPAC Name 1,2-diethenylbenzene; 1-ethenyl-2-ethylbenzene; ethenylbenzene
SMILES *

Specifications

Physical Form Beads
CAS Min % 42%, 50%
CAS Max % 50%, 58%
Ion Exchange Capacity ≥1.7 eq/L (H form)
Particle Size 100 to 200 mesh, ≤15 % (< 88 μm), ≤15 % (> 250 μm)
Packaging Plastic Bottle
Water Retention Capacity 50 to 58 % (H form)
Color Beige to Green/Beige to Light Brown
Quantity 2.5 kg
Solubility Information Insoluble in water
Chemical Name or Material AmberChrom™
Show More Show Less
Hazard Category Corrosion
Recommended Storage Normal conditions
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.