Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AmberChrom™, 50WX8, 50-100 mesh, H-form, Thermo Scientific™

Catalog No. AC472911000
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC472911000 100 g
AC472915000 500 g
AC472910025 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. AC472911000 Supplier Thermo Scientific Chemicals Supplier No. 472911000
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Physical Form Beads
CAS 69011-20-7, 7732-18-5
CAS Min % 0.5
CAS Max % 0.58
Color Beige to Green/Beige to Light Brown
Quantity 100 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.