Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AmberChrom™, 50WX8, 50-100 mesh, H-form, Thermo Scientific™

Catalog No. AC472915000
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC472915000 500 g
AC472911000 100 g
AC472910025 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. AC472915000 Supplier Thermo Scientific Chemicals Supplier No. 472915000
  • $401.00
    $327.65 / Each of 1

    Save $73.35

    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Physical Form Beads
Chemical Name or Material AmberChromTM
Particle Size 50 to 100 mesh; =<20% (< 180 μm); =<15 % (> 500 μm)
Packaging Plastic bottle
Water Retention Capacity 50 to 56 % (H form)
MDL Number 00132726
Color Beige to Brown
Quantity 500 g
Shipping Condition No special requirements
Product Line Accessories
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.