Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ AmberChrom™ (formerly Dowex™) 50WX2 Hydrogen Form, 200-400 mesh

Catalog No. 111015234 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5234 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5234 Supplier MilliporeSigma™ Supelco™ Supplier No. 217476500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A preferred strong cationic exchange (SCX) resin due to its attractive features such as high capacity, easy availability and ease of operation owing to the lower degree of divinylbenzene cross-linking leading to higher recoveries of aromatic amino acids.

Dowex™ 50WX2 hydrogen form is a heat stable, water stable and low molecular weight cationic substance which is capable of purifying approximately 5000-fold by ion-exchange chromatography. Ion-exchange resins have good role as drug delivery systems.
TRUSTED_SUSTAINABILITY

Specifications

Particle Size 200/400 Mesh
Packaging Poly Bottle
pH 0 to 14
Quantity 500 g
Product Line Amberchrom
Capacity (Metric) 0.6 meq/mL by wetted bed volume
MDL Number MFCD00132722
CAS 69011-20-7

Amberchrom is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.