Learn More
GFS Chemicals Inc AMBERLITE™ IRC120 H, ION-EXCHANGE RESIN | 500 G | GFS CHEMICALS, INC. Item #: 11822 | Bulk #: 2642
SDP
Catalog No. NC1669258
Encompass_Preferred
AMBERLITE™ IRC120 H, ION-EXCHANGE RESIN | Chemical Formula : | CAS # : 39389-20-3 | Specifications : Assay (Whole Beads) (%) 95 % - Min, Particle Size <300 micron (%) 2.0 % , Particle Size >1180 micron (%) 3.0 %
Catalog No. NC1669258 Supplier GFS Chemicals Inc Supplier No. 11822
AMBERLITE™ IRC120 H, ION-EXCHANGE RESIN | Chemical Formula : | CAS # : 39389-20-3 | Specifications : Assay (Whole Beads) (%) 95 % - Min, Particle Size <300 micron (%) 2.0 % , Particle Size >1180 micron (%) 3.0 %