Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amberlite 200, MP Biomedicals™

Catalog No. ICN15027880 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15027880 100 g
ICN15027890 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15027880 Supplier MP Biomedicals, Inc Supplier No. 0215027880
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Strongly acidic macroreticular-type cation exchange resin. Sodium form sulfonic acid functionality

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12626-25-4
MDL Number MFCD00145784

Specifications

Physical Form Powder
Color Yellow
Quantity 100 g
Chemical Name or Material Amberlite
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.