Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Amberlite™ FPA66 (formerly Dowex 66), Free Base

Catalog No. p-7191696 Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
1 kg

Weakly basic anion exchange resin for deashing and mixed bed polishing of high fructose corn syrups. This resin meets requirements of FDA Food Additive Regulation 21 CFR 173.25.

This resin has not been specially processed nor cleaned. It is suggested that it be subjected to a suitable preliminary elution and wash.Specifications:
TRUSTED_SUSTAINABILITY

Specifications

Packaging Fiber Drum
pH 0 to 7
Quantity 1 kg
Product Line Amberlite
Capacity (Metric) 1.35 meq/mL by wetted bed volume
MDL Number MFCD00135579

Amberlite is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.