Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Amberlite™ FPC23, Hydrogen Form, 16-40 mesh

Catalog No. 111014114 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4114 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4114 Supplier MilliporeSigma™ Supelco™ Supplier No. 413491KG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Amberlite™ FPC23 is proton ion-exchanger resin. Amberlite™ FPC23 H, macroreticular strongly acidic cation exchange resin for deashing of corn syrups and the softening of beet sugar syrups.

  • Amberlite™ FPC23 is a macroreticular, strongly acidic cation exchange resin designed for applications such as the deashing of corn syrups and the softening of beet sugar syrups.
  • It is also used to obtain protonated nanostructured aluminosilicate via ion-exchange columns.
  • Material: Styrene-divinylbenzene
  • Form: Beads
  • Particle Size: 16-40 mesh
  • Mean Pore Size: 200 Å
  • Surface Area: 800 m²/g
  • Technique: Suitable for LPLC
  • Loss on Drying: 40 to 55%
TRUSTED_SUSTAINABILITY

Specifications

Particle Size 16/40 Mesh
Pore Size 200 Å
Technique Chromatography
Quantity 1 kg
Surface Area (Metric) 800 m2/g
Product Line Amberlite
Material Styrene-divinylbenzene
CAS 39389-20-3

Amberlite is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.