Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amberlite™ IRA-402, Cl-form, ion-exchange resin

Catalog No. p-4259095
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC369942500 250 g Plastic bottle
AC369940010 1 kg Plastic bottle
AC369940050 5 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC369942500 Supplier Thermo Scientific Chemicals Supplier No. 369942500
Only null left
Add to Cart

CAS: 52439-77-7

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Ion exchange resin
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52439-77-7
MDL Number MFCD00145564

Specifications

Physical Form Beads
Particle Size Particle size analysis: uniformity coefficient: <1.60, harmonic mean size: 0.600 to 0.750mm,% <0.300mm: <1.0%,%>1.180mm: <5.0%
Packaging Plastic bottle
Volume Total Capacity (Cl):> 1.20 eq/L
Water Retention Capacity (Cl): 49.0 to 60.0%
Color Yellow
Quantity 250 g
Solubility Information Solubility in water: insoluble.
Chemical Name or Material Amberlite™ IRA-402

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.