Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amberlite IRA-900, MP Biomedicals™

Catalog No. ICN15031991 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15031991 1 kg
ICN15031980 100 g
ICN15031990 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15031991 Supplier MP Biomedicals, Inc Supplier No. 0215031991
Add to Cart
Add to Cart

Chemical Identifiers

CAS 9050-97-9
Molecular Formula C4H12ClN
Molecular Weight (g/mol) 109.60
MDL Number MFCD00132712
InChI Key OKIZCWYLBDKLSU-UHFFFAOYSA-M
PubChem CID 6379
ChEBI CHEBI:7070
IUPAC Name tetramethylazanium chloride
SMILES [Cl-].C[N+](C)(C)C

Specifications

Physical Form Solid
Quantity 1 kg
Chemical Name or Material Amberlite IRA-900
Recommended Storage Store at 2° to 8°C.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.