Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Amberlite IRA-938, ion-exchange resin, Thermo Scientific™
Click to view available options
Quantity:
100g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
CAS | 52932-61-3 |
MDL Number | MFCD00081326 |
Merck Index | 14,382 |
Quantity | 100g |
Chemical Name or Material | Amberlite IRA-938, ion exchange resin |
Safety and Handling
Hazard Category | Warning |
Hazard Statement | GHS H Statement H319 Causes serious eye irritation. |
Precautionary Statement | GHS P Statement P280-P264-P305+P351+P338-P337+P313 Wear protective gloves/protective clothing/eye protection/face protection. Wash thoroughly after handling. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF eye irritation persists: Get medical advice/attention. |
TSCA | TSCA |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction