Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amberlite™ IRA-958(Cl), ion exchange resin

Catalog No. AA42702A1
Change view
Click to view available options
Quantity:
1 kg
5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42702A1 1 kg
AA42702A7 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA42702A1 Supplier Thermo Scientific Chemicals Supplier No. 042702A1

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Removes Color, organics, and Cyanide

Removal of color, organics, and cyanideAmberlite™ IRA-958(Cl), ion exchange resin is used as an organic scavenger for the adsorption of natural organic matter from surface water. It is an acrylic resin for decolorization and the removal of organic material. Further, it is useful for decolorizing sugar juices and cane syrups, citric acid purification and capturing metal complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Physical Form Beads
Chemical Name or Material Amberlite IRA-958(Cl)
MDL Number MFCD00211600
Color White
Quantity 1 kg
Solubility Information Insoluble in water.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.