Learn More
Description
- Amberlite™ IRA-96 is an industrial grade, weakly basic anion exchange resin consisting of spherical beads of a macroporous styrene-divinylbenzene copolymer with a polyamine functional group.
- Functional Group: Tertiary amine (at least 85%)
- Matrix: Macroporous styrene-divinylbenzene
- Operating pH: 0 to 7
- Maximum Operating Temperature: 60°C
- Total Exchange Capacity: 1.25 eq/L
- Ionic Form as Shipped: Free base
- Particle Size: 0.550 to 0.750 mm (Harmonic mean size)
- Uniformity Coefficient: ≤1.80
- Moisture Holding Capacity: 57% to 63%
- Technique: Suitable for LPLC
Specifications
Specifications
| Technique | Chromatography |
| pH | 0 to 7 |
| Quantity | 1 kg |
| Product Line | Amberlite |
| Capacity (Metric) | 1.25 meq/mL by wetted bed volume |
| MDL Number | MFCD00166430 |
| CAS | 39409-19-3 |
Amberlite is a registered trademark of The Dow Chemical Company or an affiliated company of Dow
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.