Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Amberlite™ IRA-96, Free Base

Catalog No. p-7191674 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4128 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4128 Supplier MilliporeSigma™ Supelco™ Supplier No. 064411KG
Only null left
Add to Cart
Add to Cart

Weakly basic

TRUSTED_SUSTAINABILITY

Specifications

pH 0 to 7
Quantity 1 kg
Product Line Amberlite
Capacity (Metric) 1.25 meq/mL by wetted bed volume
MDL Number MFCD00166430
CAS 39409-19-3

Amberlite is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.