Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amberlite™ IRC-748, ion exchange resin

Catalog No. AAL195700B
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL195700B 1000 g
AAL1957030 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL195700B Supplier Thermo Scientific Chemicals Supplier No. L195700B
  • $180.00
    $166.65 / Each of 1

    Save $13.35

    7% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 79620-28-3

Amberlite™ IRC-748, ion exchange resin is used as a chelating resin with high affinity for heavy metal cations compared to alkali or alkaline earth metals. It is also utilized in metal-chelation applications. Further, it is used in purification of brine in the chloralkali industry as well as the purification of process streams having trace metals. In addition to this, it is used in the electroplating industry and in the recovery of heavy metals from hydrometallurgical leach streams.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Amberlite|r IRC-748, ion exchange resin is used as a chelating resin with high affinity for heavy metal cations compared to alkali or alkaline earth metals. It is also utilized in metal-chelation applications. Further, it is used in purification of brine in the chloralkali industry as well as the purification of process streams having trace metals. In addition to this, it is used in the electroplating industry and in the recovery of heavy metals from hydrometallurgical leach streams.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79620-28-3
MDL Number MFCD00132702

Specifications

Odor Odorless
Merck Index 14,382
Quantity 1000 g
Solubility Information Insoluble in water.
Chemical Name or Material Amberlite™ IRC-748, ion exchange resin
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.