Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Amberlite™ IRC120 H, Hydrogen Form (previously Amberlite™ IR120 H)

Catalog No. 111014127 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4127 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4127 Supplier MilliporeSigma™ Supelco™ Supplier No. 10322
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Strongly acidic cation exchange resin suitable for a wide variety of chemical process applications, removal of amino acids at low pH, USP potassium methods, etc.

Amberlite™ IR 120-H, gel type acidic cation exchange resin is of sulfonated polystyrene type having tremendous stability. It is an efficient heterogeneous catalyst for organic reactions.
TRUSTED_SUSTAINABILITY

Specifications

Packaging Bucket
pH 0 to 14
Quantity 500 g
Product Line Amberlite
Capacity (Metric) 1.8 meq/mL by wetted bed volume
MDL Number MFCD00132707
CAS 39389-20-3

Amberlite is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.