Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Amberlite IRN150, Hydrogen and Hydroxide Form

Catalog No. 111001882 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-1882 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-1882 Supplier MilliporeSigma™ Supelco™ Supplier No. 002341KG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mixed bed resin for primary water chemistry control in once-through systems.

Useful in industrial water treatment where as-supplied resin must have absolute minimum of ionic and non-ionic contamination.
TRUSTED_SUSTAINABILITY

Specifications

Grade Nuclear Grade
Quantity 1 kg
Product Line Amberlite
Description Strongly basic, 60°C max. temp., 8% cross link

Amberlite is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.