Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Amberlite™ IRN78 OH, Hydroxide Form

Catalog No. 111002405 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-2405 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-2405 Supplier MilliporeSigma™ Supelco™ Supplier No. 10343U
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Amberlite™ IRN78 hydroxide form was used as packing bed in vertical column chromatography.

Amberlite™ IRN78, strong base anion exchange gel type polystyrene resin in hydroxide form, has quaternary ammonium functional group.
TRUSTED_SUSTAINABILITY

Specifications

Grade Nuclear Grade
Packaging Bucket
pH 0 to 14
Quantity 500 g
Product Line Amberlite
Capacity (Metric) ≥1.1 meq/mL by wetted bed volume
CAS 11128-95-3

Amberlite is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.