Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amberlite™ MAC-3 H, Hydrogen Form, MilliporeSigma™ Supelco™

Catalog No. 111015870 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5870 250 g
11-101-5871 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-5870 Supplier MilliporeSigma Supelco Supplier No. 546976250G
Add to Cart
Add to Cart

Dowex MAC-3 contains carboxylic acid as functional group and is selective for Cu and Fe. The resin has larger exchange capacity and large particle size.

TRUSTED_SUSTAINABILITY

Specifications

CAS 9052-45-3
Product Line Amberlite
MDL Number MFCD03098973
Type Organic Standard
Quantity 250 g

Amberlite is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.