Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ AmberLite MB20 H/OH Mixed Bed Ion Exchange Resin, Hydrogen and Hydroxide Form

Catalog No. 111002507 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1000 g
Packaging:
Bucket
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
11-100-2507 1000 g Bucket
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-2507 Supplier MilliporeSigma™ Supelco™ Supplier No. 13687U

missing translation for 'validMainframeMsg'

Formerly Dowex™ Marathon™ MR-3 H/OH

Dowex Marathon MR-3 is a premium, premixed, mixed bed product designed for polishing demineralized water to conductivities < 0.1 muS/cm. Marathon MR-3 is a 1:1 equivalent mixture of Marathon C-10 (H) and Marathon A (OH)
TRUSTED_SUSTAINABILITY

Specifications

Packaging Bucket
pH 0 to 14
Quantity 1000 g
Product Line Marathon
Capacity (Metric) 1.0 meq/mL by wetted bed volume (anion)
MDL Number MFCD00166706

Marathon is a trademark of The Dow Chemical Company or an affiliated company of Dow

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.