Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Amberlite™ XAD-2

Catalog No. 111016353 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 g
500 g
5 kg
10 kg
25 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-6353 100 g
11-101-6350 500 g
11-100-6584 5 kg
11-100-1832 10 kg
11-100-1830 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 11-101-6353 Supplier MilliporeSigma™ Supelco™ Supplier No. 20275
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Amberlite™ XAD™-2 was used: as adsorbent in the extraction of organochlorine compounds using Soxhlat extraction technique, as adsorbent in the extraction of honey samples using column chromatography for HPLC analysis and in adsorption chromatography

Nonionic macroreticular resin that adsorbs and releases ionic species through hydrophobic and polar interactions; usually used under isocratic conditions.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 100 g
Surface Area (Metric) ˜300 m2/g
Product Line Amberlite
Density 1.02 g/mL at 25°C (true wet)(lit.)
CAS 9003-70-7

Amberlite is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.