Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amberlite™ XAD-4

Catalog No. p-2885562
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC202231000 100 g Plastic bottle
AC202235000 500 g Plastic bottle
AC202230025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC202231000 Supplier Thermo Scientific Chemicals Supplier No. 202231000
Only null left
Add to Cart

CAS: 9003-69-4 | C10H10

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Ion exchange resin
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9003-69-4
Molecular Formula C10H10
Molecular Weight (g/mol) 0.00
MDL Number MFCD00132704
InChI Key MYRTYDVEIRVNKP-UHFFFAOYSA-N
Synonym divinylbenzene, 1,2-divinylbenzene, o-divinylbenzene, benzene, diethenyl, benzene, 1,2-diethenyl, divinylbenzen, divinyl benzen, divinyl-benzene, poly divinylbenzene, divinylbenzene dvb
PubChem CID 66666
IUPAC Name 1,2-diethenylbenzene
SMILES *

Specifications

Physical Form Crystalline Powder or Crystals
Particle Size 2.0 (uniformity coefficient) Max. 0.490 to 0.690mm (harmonic mean size) 5.0% Max. (<0.300mm) 5.0% Max. (>1.180 mm)
Packaging Plastic bottle
Water Retention Capacity moisture holding capacity: 54.0 to 60.0%
Color White
Quantity 100 g
Chemical Name or Material Amberlite™ XAD-4

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.