Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amberlite XAD-7

Catalog No. AAL1956422
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1956422 100 g
AAL1956436 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1956422 Supplier Thermo Scientific Chemicals Supplier No. L1956422
Add to Cart

CAS: 37380-43-1

Amberlite XAD-7 is used in chromatography media, enzyme immobilization and water purification. It is also used in the removal of relatively polar compounds from non aqueous solvents, non-aromatic compounds from polar solvent, recovery of plant extracts, antibiotics, enzymes and proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Amberlite XAD-7 is used in chromatography media, enzyme immobilization and water purification. It is also used in the removal of relatively polar compounds from non aqueous solvents, non-aromatic compounds from polar solvent, recovery of plant extracts, antibiotics, enzymes and proteins.

Solubility
Insoluble in water.

Notes
Keep the container tightly sealed. Store in a cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37380-43-1
MDL Number MFCD00132705

Specifications

Merck Index 14,382
Quantity 100 g
Solubility Information Insoluble in water.
Chemical Name or Material Amberlite XAD-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.