Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Amberlite™ XAD4, 20-60 mesh

Catalog No. 111015790 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5790 500 g
11-101-5791 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-5790 Supplier MilliporeSigma™ Supelco™ Supplier No. XAD4500G
Add to Cart
Add to Cart

Hydrophobic polyaromatic resin

Nonionic macroreticular resin that adsorbs and releases ionic species through hydrophobic and polar interactions; usually used under isocratic conditions.
TRUSTED_SUSTAINABILITY

Specifications

Particle Size 20/60 Mesh
Pore Size 100 Å
Packaging Poly Bottle
Quantity 500 g
Surface Area (Metric) 750 m2/g
Product Line Amberlite
MDL Number MFCD00132704
Density 1.08 g/mL (skeletal)(lit.)
CAS 37380-42-0

Amberlite is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.