Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Amberlite™ XAD4, 20-60 mesh

Catalog No. 111016352 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-6352 100 g
11-101-6349 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-6352 Supplier MilliporeSigma™ Supelco™ Supplier No. 20276
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Two particle sizes are available: Mesh 20-60 are Cat. Nos. 20276, 10358, XAD4 Mesh 20-50 is Cat. No. 06444

Amberlite™ XAD4, a nonpolar absorbent, used for stripping and concentration of organic compounds. It is also used in developing chelating ligands due to its properties like, porosity, high surface area, purity and durability.
TRUSTED_SUSTAINABILITY

Specifications

Particle Size 20/60 Mesh
Pore Size 100 Å
Quantity 100 g
Surface Area (Metric) 750 m2/g
Product Line Amberlite
MDL Number MFCD00132704
Density 1.08 g/mL (skeletal)(lit.)
CAS 37380-42-0

Amberlite is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.