Learn More
MilliporeSigma™ Supelco™ Amberlite™ XAD7HP, 20-60 mesh
Catalog No. 111005089 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 g
Catalog No. Quantity
11-100-5089 100 g
1 options
Catalog No. 11-100-5089 Supplier MilliporeSigma™ Supelco™ Supplier No. XAD7100G
Add to Cart
Add to Cart
Moderately polar acrylic resin
Nonionic macroreticular resin that adsorbs and releases ionic species through hydrophobic and polar interactions; usually used under isocratic conditions.
TRUSTED_SUSTAINABILITY
Specifications
Particle Size 20/60 Mesh
Packaging Poly Bottle
Quantity 100 g
Surface Area (Metric) 380 m2/g
Product Line Amberlite
MDL Number MFCD00132705
Amberlite is a registered trademark of The Dow Chemical Company or an affiliated company of Dow