Learn More
Sigma Aldrich Amberlite™ Hpr1100
Catalog No. 919731KGF Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
91-973-1KGF 250 mg
91-973-250GF 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 91-973-1KGF Supplier Sigma Aldrich Supplier No. 919731KGF
Only null left
Add to Cart
Add to Cart
Warning
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
AMBERLITE™ HPR1100 is a cation-exchange resin and can be used:For grafting polyethylene glycols for catalyzing regioselective azidolysis of epoxides.For the immobilization of +a-cyclodextrin to be used as a phase transfer catalyst for the reduction of epoxides to alcohols.As an adsorbent for the removal of arsenic and copper from aqueous solutions.Amberlite is a registered trademark of The Dow Chemical Company or an affiliated company of Dow
Specifications
Quantity 250 mg