Learn More
Description
Specifications
Specifications
| Particle Size | 0.10 to 0.3 mm |
| Technique | Chromatography |
| Packaging | Poly Bottle |
| pH | 0 to 14 |
| Quantity | 500 g |
| Product Line | Amberlyst |
| Description | Swelling (dry to solvent saturated, %):hexane 10-15; toluene 10-15; ethylene dichloride 15-20; ethylene acetate 30-40; 95% ethylene alcohol 60-70; water 60-70. |
| Material | Styrene-divinylbenzene |
| Capacity (Metric) | 4.7 meq/g by dry weight |
| MDL Number | MFCD00145841 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.