Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Amberlyst™ 15, Hydrogen Form, Wet

Catalog No. 111014119 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4119 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4119 Supplier MilliporeSigma™ Supelco™ Supplier No. 216399500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A macro reticular polystyrene based ion exchange resin with strongly acidic sulfonic group.

Amberlyst 15 serves as an excellent source of strong acid. It can also be used several times. Amberlyst 15 is a heat sensitive macro-porous sulfonic ion exchange acid resin which could be used as a catalyst.
TRUSTED_SUSTAINABILITY

Specifications

Particle Size 0.5 to 1.0 mm
Technique Chromatography
Packaging Poly Bottle
pH 0 to 14
Quantity 500 g
Product Line Amberlyst
Description Swelling (dry to solvent saturated, %):hexane 10-15; toluene 10-15; ethylene dichloride 15-20; ethylene acetate 30-40; 95% ethylene alcohol 60-70; water 60-70.
Material Styrene-divinylbenzene
Capacity (Metric) 4.7 meq/g by dry weight
MDL Number MFCD00145841
CAS 39389-20-3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.