Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Amberlyst™ 36

Catalog No. 111014126 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4126 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4126 Supplier MilliporeSigma™ Supelco™ Supplier No. 4367121KG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

51-57% Moisture content (H+ form)

Shrinkage: Water to phenol: 45%; Water to dry: 54%
TRUSTED_SUSTAINABILITY

Specifications

Pore Size 240 Å
Packaging Poly Bottle
Quantity 1 kg
Surface Area (Metric) 33 m2/g
Product Line Amberlyst
Description Water, 55%
Capacity (Metric) >1.95 eq/L exchange capacity
MDL Number MFCD00145841
Density 1.2 g/mL at 25°C (lit.)
CAS 39389-20-3

Amberlyst is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.