Learn More
Description
Specifications
Specifications
| Technique | Chromatography |
| Packaging | Poly Bottle |
| Quantity | 250 g |
| Product Line | Amberlyst |
| Material | Styrene-divinylbenzene |
| Capacity (Metric) | 1.3 meq/mL by wetted bed volume |
| MDL Number | MFCD00145842 |
Amberlyst is a registered trademark of The Dow Chemical Company or an affiliated company of Dow
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.