Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amberlyst™ A26 Hydroxide Form, MilliporeSigma™ Supelco™

Catalog No. 111014113 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4113 1 kg
11-101-4112 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-4113 Supplier MilliporeSigma Supelco Supplier No. 5425711KG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Inert in strong acids, concentrated aldalies, aliphatic and aromatic hydrocarbons, alcohols, ethers, and other common solvents.Maximum operating temperature 60C.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Styrene-divinylbenzene
CAS 39339-85-0
Product Line Amberlyst
Physical Form Beads
Concentration 1 N in NaOH (Regenerant concentration)
Technique Ion Chromatography
MDL Number MFCD03096474
Type Organic Standard
Quantity 1 kg
Grade Analytical

Amberlyst is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.