Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amberlyst™ A26, OH-form, ion-exchange resin

Catalog No. AC202180010
Change view
Click to view available options
Quantity:
100 mL
1 L
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC202180010 1 L Plastic bottle
AC202181000 100 mL Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC202180010 Supplier Thermo Scientific Chemicals Supplier No. 202180010
Only null left
Add to Cart

CAS: 39339-85-0

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Ion exchange resin
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39339-85-0
MDL Number MFCD00145579

Specifications

Packaging Plastic bottle
Density 0.6750g/mL
Fieser 01,511; 02,227; 04,266; 05,355; 06,302; 07,182; 08,263
Merck Index 10, 381
Quantity 1 L
Chemical Name or Material Amberlyst™ A26

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.