Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ambertec™ UP550 OH Anion Exchange Resin, Sold by MilliporeSigma™ Supelco™

Catalog No. 111022422 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
20 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-2422 20 kg
11-102-2420 1 kg
11-102-2421 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-102-2422 Supplier MilliporeSigma Supelco Supplier No. 14019U
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dowex™ Monosphere™ 550A UPW OH type 1 strong base anion exchange resin is recommended for the roughing, intermediate, and polishing ion exchange applications. Low TOC levels in UPW grade are achieved by deliberate functionalization and rinsing.

  • Light color
  • Superior crush strength
  • High (>95%) conversion of exchange sites to the hydroxide (OH) form
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Formerly Dowex™ Monosphere™ 550 A UPW OH
CAS 69011-18-3, 7732-18-5
Particle Size 590 μm (Standard Deviation: ±50)
Volume Total Capacity 1 eq/L
Water Retention Capacity 55% to 65%
Quantity 20 kg
Grade UPW
Concentration or Composition (by Analyte or Components) Trimethylamine functionalized copolymer of styrene and divinylbenzene in the hydroxide form

AmberTec is a trademark of E. I. du Pont de Nemours and Company

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.