Learn More
Invitrogen™ TBE (10X), RNase-free
Description
Molecular biology grade, Invitrogen 10X TBE solution is supplied in four bottles containing 1 L each. The buffer is certified RNase-free, economical, and ready-to-use. Due to the ubiquitous presence of RNases, manufacturing products for use with RNA is especially challenging. Invitrogen's nuclease-free reagents and buffers are manufactured in facilities specifically designed to prevent the introduction of nucleases. Highly sensitive RNase assays are performed at several different stages of the manufacturing process to ensure the highest quality. These reagents are rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.
Order Info
Shipping Condition: Room temperature
Certifications
Certified RNase-free
Specifications
Specifications
| Chemical Name or Material | Running Buffer |
| Concentration | 10X |
| Physical Form | Liquid |
| Product Line | Ambion |
| Quantity | 4 x 1 L |
| Grade | Molecular Biology |
Frequently Asked Questions (FAQs)
No. Autoclaving at temperatures that are too high or pressures that are too high can hydrolyze the agarose polymer and weaken it.
Add the powder to cold buffer while stirring. Let the agarose rehydrate for 1 to 2 hr and then heat slowly until agarose is completely dissolved. Using a microwave at low power settings is acceptable.
One recommendation: get a supply of fabric softener sheets and wipe the bottle with a sheet before opening it.
The precipitation of concentrated TBE stocks may be due to nucleation of salt crystals by dust particles or other insoluble materials. Therefore, filtering the solution through a 0.2 µm cellulose acetate or cellulose nitrate filter after preparation helps avoid this precipitate. [Mayeda A, Krainer A (1991) BioTechniques 10.2, 1820]
Although there is only a very small chance of creating replication competent virus, steps should still be taken to avoid added risk. The virus is expanded in two rounds of amplification, and all secondary expansions should be performed from an initial expansion stock. Secondary amplifications in HEK293 cells should not be performed from the stock of another secondary expansion, as this could increase the chance of making replication competent virus. See the product manual for more detail and guidelines for screening.
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.