Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cap Analog (m7G(5')ppp(5')G), 10 A254 units

Catalog No. AM8048
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Tube
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM8048 1 Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM8048 Supplier Invitrogen™ Supplier No. AM8048
Season of Savings
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Single tube (10 A254 U).

For 5' capped RNA molecules synthesis as part of in vitro transcription reactions

Invitrogen Cap Analog [m7G(5')ppp(5')G] is used for the synthesis of 5' capped RNA molecules as part of in vitro transcription reactions. These molecules are translated more efficiently using in vitro translation systems than uncapped mRNAs. 10 A254 U supplied.

  • Used as a highly specific inhibitor of the initiation step of protein synthesis.
  • Tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity
  • Assayed for efficient synthesis of capped in vitro transcripts by using cap analog in functional MEGAscript™ In Vitro transcription and Retic Lysate translation reactions

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Supplied in a single tube to be stored at or below –70°C. Avoid multiple freeze-thaw cycles. Aliquots of the product may be stored short-term at –20°C.
Concentration 40 mM
Final Product Type Cap Analog for In vitro Transcription, Cap Analog for mRNA
For Use With (Application) In Vitro Transcription, synthetic mRNA, mRNA
For Use With (Equipment) mMessage mMAchine, MEGAscript
Labeling Method RNA Labeling
Product Line Ambion
Product Type Cap analog for mRNA, cap analog for synthetic mRNA
Quantity 1 Tube
Shipping Condition Dry Ice
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.