Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ dNTP Mix (2.5 mM)

Catalog No. AM8228G
Encompass_Preferred
Change view
Click to view available options
Quantity:
300 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM8228G 300 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM8228G Supplier Invitrogen™ Supplier No. AM8228G
  • $206.00
    $197.65 / Each of 1

    Save $8.35

    4% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

The same high-quality deoxynucleotide triphosphate solutions provided in the Invitrogen™ RETROscript™ First Strand cDNA Synthesis Kit for RT-PCR.

The same high-quality deoxynucleotide triphosphate solutions provided in the Invitrogen™ RETROscript™ First Strand cDNA Synthesis Kit for RT-PCR (Cat. No. AM1710). They are subject to rigorous nuclease testing as well as a functional assay to ensure high levels of incorporation. Supplied in one tube containing 0.3 mL, premixed at a concentration of 2.5 mM each.

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at –20°C.
Concentration 2.5 mM
For Use With (Application) Reverse Transcription
Label or Dye Unlabeled
Product Line Ambion
Quantity 300 μL
Shipping Condition Dry Ice
Why do some dNTP mixes contain dTTP and others TTP?

Thymine is normally found as a deoxyribose. Thus, TTP and dTTP are essentially the same thing. Both types of mixes can be used in PCR reactions

WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.