Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Gel Loading Solution (All-Purpose, Native Agarose)

Catalog No. AM8556
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM8556 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM8556 Supplier Invitrogen™ Supplier No. AM8556
Get 19% Off Invitrogen Molecular Biology Reagents
Use promo code INV19
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

A 10X solution of 40% sucrose, 0.17% xylene cyanol, and 0.17% bromophenol blue

  • Rigorously tested for non-specific endonuclease activity, exonuclease activity, RNase activity and for functionality
  • Compatible with native agarose and polyacrylamide gels
  • Bromophenol Blue and Xylene Cyanol dyes
  • Supplied in one 1.4mL tube
  • The same solution in Invitrogen kits

Acrylamide Gel Electrophoresis, Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Northern Blotting, Nucleic Acid Gel Electrophoresis and Blotting

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at -20°C.
Buffer Sample Loading Buffers
Concentration 10X
Label or Dye Bromophenol Blue, Xylene Cyanol
Gel Compatibility Polyacrylamide Gels, Native Agarose
Quantity 1 mL
Shipping Condition Room Temperature
Product Type Gel Loading Solution
How can generation of replication competent adenoviruses be avoided when using your pSilencer adeno 1.0-CMV System?

Although there is only a very small chance of creating replication competent virus, steps should still be taken to avoid added risk. The virus is expanded in two rounds of amplification, and all secondary expansions should be performed from an initial expansion stock. Secondary amplifications in HEK293 cells should not be performed from the stock of another secondary expansion, as this could increase the chance of making replication competent virus. See the product manual for more detail and guidelines for screening.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.