Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Lung Total RNA

Catalog No. AM7968
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM7968 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM7968 Supplier Invitrogen™ Supplier No. AM7968
Season of Savings
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The highest-quality RNA available from Invitrogen, it is DNase-treated, subjected to unsurpassed quality control standards, and certified to contain small RNAs (miRNA, siRNA, and snRNA). One tube containing 100 μg is provided at a concentration of 1 mg/mL.

The highest-quality RNA available from Invitrogen, it is DNase-treated, subjected to unsurpassed quality control standards, and certified to contain small RNAs (miRNA, siRNA, and snRNA). One tube containing 100 μg is provided at a concentration of 1 mg/mL. Intactness of mRNA, genomic DNA contamination, presence/absence of enzymatic inhibitors, and ribosomal RNA contamination in poly(A) preparations are all important aspects used to judge the quality of FirstChoice™ RNAs. The samples are processed using Invitrogen RNA isolation reagents to produce highly pure, intact RNA. A stringent DNase treatment is performed to ensure that the RNA is ready for use in any downstream application, including RT-PCR. Because it is impossible to remove every DNA molecule from an RNA sample, each preparation is tested to ensure that any residual DNA contamination is insignificant. The integrity of the RNA is verified by capillary electrophoresis using the Agilent™ 2100 bioanalyzer.

Order Info

Shipping Condition: Dry ice

Certifications

microRNA-Certified

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 mg/mL
Content And Storage Store at or below -70°C. Avoid multiple freeze–thaw cycles. Aliquots of the product may be stored short-term at -20°C. DO NOT STORE IN FROST-FREE FREEZER.
Species Human
Sample Type Total RNA
Product Line Ambion
Purity microRNA-Certified
Quantity 100 μg
Sample Treatment DNase-Treated
Sample Source Normal Tissue
Shipping Condition Dry Ice
Tissue Lung
Show More Show Less
Do you offer isolated, purified RNA?

Yes, please visit this website (http://www.thermofisher.com/us/en/home/life-science/dna-rna-purification-analysis/rna-extraction/rna-types/purified-rna.html) for our FirstChoice total RNA isolated from human tissues, as well as from E. coli and human cell lines.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.