Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MEGAshortscript™ T7 Transcription Kit

Catalog No. AM1354
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM1354 25 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM1354 Supplier Invitrogen™ Supplier No. AM1354
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Maximize the yield of short RNA transcripts

The MEGAshortscript T7 Kit is designed for high yields of in vitro-transcribed RNA products, in the 20 to 500 nucleotide range. Using conventional methods, transcription of small templates often results in low mass yields of RNA. Short transcripts require many more transcription initiation events to synthesize a given mass of product. Since the initiation step is the rate-limiting step in transcription reactions, the yield of product from short templates is often low. The MEGAshortscript T7 Kit has been designed to overcome the drawbacks involved in conventional in vitro transcription to maximize the yield of short RNA transcripts.

  • Efficiently transcribes small templates (<300 bases) with consistently high yields
  • Designed for use with plasmid, synthetic oligonucleotide, or PCR product templates
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage T7 Enzyme Mix, 10X Reaction Buffer, ATP Solution, CTP Solution, GTP Solution, UTP Solution, pTRI-RNA 18S, TURBO DNase, Ammonium Acetate Stop Solution, and Gel Loading Buffer II are all stored at –20°C. Nuclease-free Water may be stored at any temperature.
Final Product Type Synthetic mRNA, mRNA
Format Kit
Sample Type mRNA, Synthetic mRNA
For Use With (Application) In Vitro Transcription, synthetic mRNA, mRNA, transfection, LNPs, GENEart, linearized DNA plasmid, delivery, PCR products
No. of Reactions 25 Reactions
Product Line Ambion, MEGAshortscript
Product Type T7 In Vitro Transcription kit
Quantity 25 Reactions
Promoter T7
Shipping Condition Dry Ice
Starting Material PCR Products, Linearized DNA plasmid
Show More Show Less
What size template should I use with the MEGAshortscript T7 kit?

The MEGAshortscript Kit is designed to function best with templates that code for RNA transcripts in the 20 to 500 nt range. The kit can be used to produce longer RNA but the Invitrogen MEGAscript Kit is more economical for this purpose.

I am planning on ordering Invitrogen GeneArt CRISPR T7 Strings DNA. What do you recommend I use to in vitro transcribe gRNA?

We recommend using our Invitrogen Megashortscript T7 Transcription Kit in order to produce ready-to-use in vitro transcribed gRNA.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.