Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ mMESSAGE mMACHINE™ T7 ULTRA Transcription Kit

Catalog No. AM1345
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM1345 10 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM1345 Supplier Invitrogen™ Supplier No. AM1345
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

The mMESSAGE mMACHINE T7 ULTRA Transcription Kit generates RNA transcripts with higher protein yields due to the combination of the anti-reverse cap analog (ARCA) with patented transcription reagents.

The mMESSAGE mMACHINE T7 Ultra Kit combines the anti-reverse cap analog (ARCA) with a patented high-yield transcription technology to generate RNA transcripts that produce higher protein yields compared to other transcripts upon translation.

Features of the mMESSAGE mMACHINE T7 Ultra Kit include:

  • Synthesize more protein from in vitro-transcribed RNA
  • Express RNA transcripts more efficiently both in vitro and in vivo
  • Stabilize transcripts in vivo with included reagents for poly(A) tailing

ARCA-capped RNA is translationally more active

A base modification in ARCA results in its incorporation in the functional, translatable orientation only; traditional cap analog can be incorporated in both functional and nonfunctional orientations. As a result, incorporating ARCA into transcription reactions yields capped RNAs that are 100% translatable. This is further enhanced by the inclusion of poly(A) tails in mMESSAGE mMACHINE T7 Ultra transcripts. Experiments comparing ARCA and ARCA/poly(A)-tailed transcripts to cap analog and cap analog/poly(A)-tailed transcripts indicate higher levels of protein synthesis with ARCA capped RNA.

What Is ARCA?

ARCA is a modified cap analog in which the 3' OH group (closer to m7G) is replaced with OCH3 (see schematic). Because of this substitution, the RNA polymerase can only initiate transcription with the remaining hydroxyl group, thus forcing ARCA incorporation in the forward orientation. As a result, 100% of the transcripts synthesized with ARCA at the 5' end are translatable, leading to a strong stimulatory effect on translation.

Proper capping of in vitro transcribed RNA

Proper capping of RNA promotes correct initiation of protein synthesis, as well as stability and processing of mRNA in vivo. Uncapped RNA is rapidly degraded by cellular RNases after microinjection or transfection into cells. Capped RNA is also typically translated more efficiently in in vitro translation systems, generating RNAs with cap analog incorporated only in the functional orientation. Therefore, substitution of traditional cap analog with ARCA results in the synthesis of capped RNAs that are 100% translatable.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage T7 Enzyme Mix, 10X Reaction Buffer 2XNTP⁄ARCA Solution, GTP Solution, pTRI-Xef, TURBO DNase, Ammonium Acetate Stop Solution, Lithium Chloride Precipitation Solution, E-PAP, 5X E-PAP Buffer, ATP Solution, MnCl2, and Formaldehyde Load Dye are all stored at –20°C. Nuclease-free Water may be stored at any temperature.
Final Product Type Synthetic mRNA, mRNA, ARCA Capped mRNA
Format Kit
Sample Type mRNA, Synthetic mRNA
For Use With (Application) In Vitro Transcription, synthetic mRNA, mRNA, transfection, LNPs, GENEart, linearized DNA plasmid, delivery, PCR products
No. of Reactions 10 Reactions
Product Line Ambion, mMESSAGE mMACHINE
Product Type T7 In Vitro Transcription kit
Quantity 10 Reactions
Promoter T7
Shipping Condition Dry Ice
Yield 20-30 μg
Starting Material PCR Products, Linearized DNA plasmid
Show More Show Less
Does the Lipofectamine MessengerMAX Transfection Reagent kit include reagents for in vitro transcription of mRNA?

Although it does not, we recommend the mMESSAGE mMACHINE T7 ULTRA Transcription Kit (Cat. No. AMB1345) to generate capped and tailed mRNA for highest mRNA stability.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.