Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PureLink™ Plant RNA Reagent

Catalog No. 12322012
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-322-012 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-322-012 Supplier Invitrogen™ Supplier No. 12322012
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Supplied as a ready-to-use solution.

Plant RNA Reagent is a single-component solution optimized for the isolation of high yields of high-quality total RNA from a variety of plant tissues, especially those rich in polyphenolics (e.g., conifer needles) or starch (e.g., potato tuber or seeds).

Plant RNA Reagent is a single-component solution optimized for the isolation of high yields of high-quality total RNA from a variety of plant tissues, especially those rich in polyphenolics (e.g., conifer needles) or starch (e.g., potato tuber or seeds).

Improved quality and purity of plant RNA
Plant RNA Reagent is optimized to have less sample DNA contamination, to be twice as effective as TRIzol Reagent, and have a simplified RNA extraction protocol for minimized time and labor. The end result is high-yield, high-purity RNA (determined by A260/A280 and gel analysis) from a variety of plant material sources.

Works for milligram to gram quantities of starting material
Plant RNA Reagent provides greater flexibility for sample size and processing formats, and can be used for both small-scale (up to 0.1 g plant tissue) and large-scale (0.1–5 g plant tissue) total RNA isolation. The amount supplied (100 mL) is sufficient for 200 reactions using 100 mg of tissue, or 4 reactions using 5g of tissue.

  • Isolate high-quality RNA from difficult plant materials
  • Use for small-scale and large-scale RNA isolations
  • Fewer resources used, less hazardous, less waste

Isolated RNA is ideal for a variety of applications and products:

  • cDNA library construction kits
  • Real-Time PCR (qPCR)
  • Northern blotting kits and reagents
  • Poly(A) selection kit RNase control reagents and assays

Not high-throughput compatible (Manual)

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage PureLink™ Plant RNA Reagent is supplied as a ready-to-use solution. Store at +4°C.
Isolation Technology Organic Extraction
Purification Time 1 hr.
Sample Type Plant Samples
Final Product Type Total RNA
High-throughput Compatibility Not High-throughput Compatible (Manual)
For Use With (Application) Reverse Transcriptase PCR (RT-PCR), cDNA Library Construction, Nuclease Protection Assays, Northern Blotting
No. of Reactions 200 Preps
Quantity 100 mL
Scale Scaleable
Starting Material Amount Up to 1 g tissue, Up to 1 x 10^7 cells
Target Total RNA
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.