Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PureLink™ RNA Micro Scale Kit

Catalog No. 12183016
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-183-016 50 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-183-016 Supplier Invitrogen™ Supplier No. 12183016
Get Up to $3,000 of qPCR Products with Purchase
View all promotions
Add to Cart
Add to Cart

PureLink™ technology combines guanidine-isothiocyanate lysis with the speed, purity, and ease-of-use of silica-membrane purification.

A simple and reliable technology for everyday RNA purification
PureLink™ technology combines guanidine-isothiocyanate lysis with the speed, purity, and ease-of-use of silica-membrane purification. The safe and easy procedure can be completed in typically less than 15 minutes with >85% recovery from the first elution. The kit can be used for clean-up of purified RNA from enzymatic reactions, desalting of RNA samples after phenol extraction or ethanol precipitation, and concentration of RNA. Purify RNA from up to 10 mg of tissue or 105 cells. Clean up and concentrate RNA amounts ranging from pg to 100 μg in volumes as small as 10 μL. Purified RNA is ready for use in downstream applications such as RT-PCR, qRT-PCR, northern blots, cDNA library construction, and nuclease protection assays.

A simple and rapid procedure
Homogenize your sample in lysis buffer, add 70% ethanol, and mix. Deposit the resulting solution into a PureLink Micro kit column with collection tube, and centrifuge for one minute. Nucleic acids bind to the PureLink silica membrane in the column, and after subsequent washes and on-column digestion with DNase, purified RNA can be eluted with RNase-free water.

  • Effective: Expect excellent RNA recovery (up to 100 μg total RNA) from small samples
  • Convenient and safe: The extraction procedure uses no phenol/chloroform, CsCl gradients, or precipitation with either LiCl or ethanol
  • Fast: Obtain high-quality total RNA, typically in just 15 minutes
  • Reliable: Obtain ready-to-use RNA suitable for any downstream application
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Box 1 (PureLink™ RNA Micro Kit):
• Lysis Buffer, 125 mL
• Wash Buffer I, 50 mL
• Wash Buffer II, 15 mL
• RNase-free Water, 15.5 mL
• PureLink™ Micro Kit Columns with collection tubes, 50 each
• Collection Tubes, 50 each
• Recovery Tubes, 50 each

Box 2 (PureLink™ DNase/Carrier RNA):
• Lyophilized PureLink™ DNase, 1500 Units
• PureLink™ On-Column DNase 2X Buffer, 1 mL
• RNase-free Water, 1 mL
• Lyophilized PureLink™ Carrier RNA, 300 μg

Store all components of Box 2 at 4°C. Store remaining components at room temperature.
Isolation Technology Silica Spin Column
Purification Time 15 min. (After sample homogenization)
Elution Volume 12–22 μL
Sample Type Cells, Tissue, Laser Capture Microdissection (LCM), Enzymatic Reactions
Final Product Type Total RNA
High-throughput Compatibility Not High-throughput Compatible (Manual)
For Use With (Application) RT-PCR, qPCR, cDNA Library Construction, NGS, Microarray Analysis, Blot Hybridization, Northern Blotting, In Vitro Translation, Nuclease Protection Assays, Nucleic Acid Labeling
Quantity 50 Preps
Scale Micro
Shipping Condition Room Temperature
Starting Material Amount Cells: ≤105
Tissue: ≤5 mg
Enzymatic Reactions: ≤1.2 mL
Yield Up to 17 μg
Show More Show Less
I am using the PureLink RNA Micro Kit, and my RNA yield is very low. Can you offer some tips?

This could be due to incomplete lysis and homogenization. Here are some recommendations:

- Ensure that 10 µL of 2-mercaptoethanol was added per 1 mL of Lysis Buffer.
- Perform all steps at room temperature unless directed otherwise.
- Decrease the amount of starting material used, or increase volume of Lysis Buffer.
- Use the proper homogenization methods according to recommendations in the sample-specific protocols.
- Cut tissue samples into smaller pieces and ensure the tissue is completely immersed in the Lysis Buffer to achieve optimal lysis.

Are the buffers (Lysis Buffer, Wash Buffer I, and Wash Buffer II) used within the PureLink RNA Mini Kit (Cat. No. 12183025) and the PureLink RNA Micro Scale Kit (Cat. No. 12183016) the same?

Yes, the compositions for Lysis Buffer, Wash Buffer I, and Wash Buffer II are the same between the PureLink RNA Mini Kit (Cat. No. 12183025) and the PureLink RNA Micro Scale Kit (Cat. No. 12183016).

WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.