Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Retic Lysate IVT™ Kit

Catalog No. AM1200M
Encompass_Preferred
Change view
Click to view available options
Product Type:
Retic Lysate
Retic Lysate with Manual
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
AM1200M Retic Lysate with Manual
AM1200 Retic Lysate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AM1200M Supplier Invitrogen™ Supplier No. AM1200M

missing translation for 'validMainframeMsg'

Includes

Retic Lysate should be stored at -80°C. 20X Translation Mix minus-Met, 20X Translation Mix minus-Leu, High Salt Mix minus-Met, Low Salt Mix minus-Met, and Capped Xef-1 RNA should be stored below -70°C. Nuclease-free Water can be stored at any temperature.

Efficiently translates high molecular weight proteins and both capped and uncapped mRNAs

The Retic Lysate IVT™ Kit efficiently translates in vitro-synthesized transcripts, poly(A) RNA, and total RNA, including difficult-to-translate RNAs. The optimized lysate results in high protein yield, biological activity, and consistent performance. A report in Nucleic Acids Research (1) found that some commercial reticulocyte lysates had poor fidelity for initiation at the appropriate AUG initiator codon. The Retic Lysate IVT Kit was tested using Kozak's constructs and it demonstrated high fidelity for initiating at the proper AUG.

  • Efficiently translates a wide variety of high molecular weight proteins and both capped and uncapped mRNAs
  • Accommodates large volume of template mRNA
How it works
The lysate has been optimized for translation by the addition of an ATP-regenerating system, hemin, and calf liver tRNA. Included in the kit is an optimized Translation Mix (-Met or -Leu) for the translation of both capped and uncapped messages along with High and Low Salt buffers. These buffers can be used to maximize protein yield and activity from difficult-to-translate sequences.

1. Kozak, M. (1990) Evaluation of the fidelity of initiation of translation in reticulocyte lysates from commercial sources. Nucleic Acids Res 18:2828.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Retic Lysate should be stored at –80°C. 20X Translation Mix minus-Met, 20X Translation Mix minus-Leu, High Salt Mix minus-Met, Low Salt Mix minus-Met, and Capped Xef-1 RNA should be stored below –70°C. Nuclease-Free water can be stored at any temperature.
Format Kit
Expressed Protein Type Water Soluble Protein
Expression Mechanism Cell-Free Expression
Expression System Cell-Free
For Use With (Application) In Vitro Transcription
No. of Reactions 60 Reactions
Product Line Ambion
Product Type Retic Lysate with Manual
Quantity 60 Reactions

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.