Learn More
Invitrogen™ RNAqueous™-4PCR Total RNA Isolation Kit
Description
Includes
600μL 10X DNase I Buffer; 45μL DNase I 2U/μL; 1mL Formaldehyde Load Dye; 100μL Linear Acrylamide 5mg/mL; 50μL Positive Control Primers; 1mL 5M Ammonium Acetate; 35mL Water for 64% Ethanol; 600mL DNase Inactivation Reagent; 30mL Lysis/Binding Solution; 25mL Wash Solution 1; 35mL Wash Solution 2/3; 3 Pestles for tissue disruption; 30 Filter Cartridges; 60 Collection Tubes; 25mL Elution Solution
The RNAqueous-4PCR Kit is used for phenol-free total RNA isolation from small tissue samples (0.5 to 75 mg) or from cells (∽100 cells to ∽1 x 107 cells), using a guanidinium-based lysis/denaturant and glass fiber filter separation technology. DNA-free™ DNA removal reagents are included in the kit, making the isolated RNA especially suitable for RT-PCR.
- DNA-free RNA for RT-PCR
- Rapid, phenol-free total RNA isolation
- Novel reagent for rapid and safe removal of DNase and divalent cations
Using the RNAqueous-4PCR Kit
The RNAqueous-4PCR Kit is designed for isolatation of RNA for applications such as RT-PCR, that would benefit from its robust genomic DNA removal capabilities. When used in conjunction with RNAlater™ Tissue Collection: RNA Stabilization Solution, most tissue samples can be stored at 4°C and then easily disrupted in standard microfuge tubes using the pestles provided in the kit. No need for snap-freezing in liquid nitrogen or grinding in a mortar and pestle.
Not high-throughput compatible (Manual)
Applications:
DNA and RNA Purification and Analysis, RNA Extraction, Total RNA Isolation, Total RNA from Animal Cells and Tissues
Specifications
Specifications
| Content And Storage | • 600 μL 10X DNase 1 Buffer; store at -20°C • 45 μL DNase 1; -20°C • 1 mL Formaldehye Load Dye; -20°C • 100 μL Linear Acrylamide; -20°C • 50 μL Positive Control Primers; -20°C • 1 mL 5M Ammonium Acetate; -20°C • 35 mL Water; 4°C • 600 μL DNase Inactivation Reagent; 4°C • 30 mL Lysis/Binding Solution; 4°C • 25 mL Wash Solution #1; 4°C • 35 mL Wash Solution 2/3 (concentrated); 4°C • 3 Pestles; room temperature • 30 Filter Cartridges; room temperature • 60 Collection Tubes; room temperature • 25 mL Elution Solution; room temperature |
| Isolation Technology | Silica Spin Column |
| Purification Time | 10 min. (After sample homogenization) |
| Elution Volume | 50–120 μL |
| Sample Type | Tissue, Cells |
| Final Product Type | Total RNA |
| High-throughput Compatibility | Not High-throughput Compatible (Manual) |
| For Use With (Application) | RT-PCR, qPCR, cDNA Library Construction, NGS, Microarray Analysis, Blot Hybridization, Northern Blotting, In Vitro Translation, Nuclease Protection Assays, Nucleic Acid Labeling |
| Quantity | 30 Preps |
| Shipping Condition | Box 1: Dry Ice, Box 2: Room Temperature |
| Show More |
Frequently Asked Questions (FAQs)
RNaqueous kits are based on a rapid, filter-based RNA isolation system that does not require phenol, chloroform, or other toxic organic chemicals. The RNAqueous Total RNA Isolation Kit is designed for sample sizes of 10-75 mg of tissue or 10^6 to 10^7 cells, whereas the RNAqueous-Midi Total RNA Isolation Kit is designed for tissue samples of up to 0.5 g or 10^9 cells. The RNAqueous-Micro Total RNA Isolation Kit is optimized for the purification of total RNA from micro-sized samples such as 10-500,000 cultured cells, as few as 10 microdissected cells, or up to 10 mg tissue. The RNAqueous-96 Total RNA Isolation Kit utilizes a 96-well plate format for high-throughput RNA isolation from 100 to 2 x 10^6 cells or 0.1 to 1.5 mg of tissue. The RNAqueous-4PCR Total RNA Isolation Kit is designed for RNA isolation from small tissue samples (0.5-75 mg) and small- to medium-sized cultured cell samples (approximately 100-10^7 cells).
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.