Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNAqueous™-96 Automated Kit

Catalog No. AM1812
Encompass_Preferred
Change view
Click to view available options
Quantity:
384 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM1812 384 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM1812 Supplier Invitrogen™ Supplier No. AM1812
Get Up to $3,000 of qPCR Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

4 Filter Plates, 4 Collection Plate with lids, 4 Culture Plates, 1 Heat Plate with lid, 100mL Nuclease-free Water, 100mL Elution Solution, 100mL Lysis/Binding Solution, 100mL Rebinding Mix, and 2 x 175mL Wash Solution Concentrate, (1.25mL DNase I (20U/μL) and 12mL DNase 1 Buffer

The Invitrogen RNAqueous-96 Automated Kit is used for automated, phenol-free total RNA isolation from 100-500,000 cells or 0.1–1 mg tissue using a guanidinium-based lysis/denaturant and glass fiber filter separation technology.

The Invitrogen RNAqueous-96 Automated Kit is used for automated, phenol-free total RNA isolation from 100-500,000 cells or 0.1–1 mg tissue using a guanidinium-based lysis/denaturant and glass fiber filter separation technology. The kit contains enough reagents for 384 (4 x 96) RNA purifications. The RNAqueous-96 Automated Kit uses an RNA-binding glass-fiber filter to provide high, reproducible yields of intact RNA. An optional on-the-filter DNase treatment can be performed to ensure removal of genomic DNA for RT-PCR applications.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 4 Filter Plates; store at room temperature
• 4 Collection Plates with lids; room temperature
• 4 Culture Plates; room temperature
• 1 Heat Plate with Lid; room temperature
• 100 mL Nuclease-free Water; room temperature
• 100 mL Elution Solution; room temperature
• 100 mL Lysis/Binding Solution; 4°C
• 100 mL Rebinding Mix; 4°C
• 2 x 175 mL Wash Solution (concentrated); 4°C
• 1.25 mL DNase 1; -20°C
• 12 mL DNase 1 Buffer; -20°C
Isolation Technology Vacuum Filter Plate (Glass Fiber Filter)
Elution Volume 105 μL
Sample Type Cells, Tissue
Final Product Type Total RNA
High-throughput Compatibility High-throughput Compatible
For Use With (Equipment) MultiPROBE™ II HT, Biomek™ 2000, Liquid Handling Systems
For Use With (Application) RT-PCR, qPCR, cDNA Library Construction, NGS, Microarray Analysis, Blot Hybridization, Northern Blotting, In Vitro Translation, Nuclease Protection Assays, Nucleic Acid Labeling
Quantity 384 Preps
Shipping Condition Box 1: Dry Ice
Box 2: Room Temperature
Starting Material Amount Cells: ≤105
Tissue: 0.1–1 mg
Show More Show Less
What are the differences in the RNAqueous kits you offer?

RNaqueous kits are based on a rapid, filter-based RNA isolation system that does not require phenol, chloroform, or other toxic organic chemicals. The RNAqueous Total RNA Isolation Kit is designed for sample sizes of 10-75 mg of tissue or 10^6 to 10^7 cells, whereas the RNAqueous-Midi Total RNA Isolation Kit is designed for tissue samples of up to 0.5 g or 10^9 cells. The RNAqueous-Micro Total RNA Isolation Kit is optimized for the purification of total RNA from micro-sized samples such as 10-500,000 cultured cells, as few as 10 microdissected cells, or up to 10 mg tissue. The RNAqueous-96 Total RNA Isolation Kit utilizes a 96-well plate format for high-throughput RNA isolation from 100 to 2 x 10^6 cells or 0.1 to 1.5 mg of tissue. The RNAqueous-4PCR Total RNA Isolation Kit is designed for RNA isolation from small tissue samples (0.5-75 mg) and small- to medium-sized cultured cell samples (approximately 100-10^7 cells).

WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.